APO455835465 Apollo Scientific Ltd

3-Carboxy-4-fluorobenzeneboronic acid

  • MDL number MFCD03095129
  • Cas Number : 872460-12-3
  • Molecular Weight : 183.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 217-219
Synonym 5-Borono-2-fluorobenzoic acid
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-carboxy-4-fluorobenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-carboxy-4-fluorobenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart