APO455835469 Apollo Scientific Ltd

1,2-Dimethoxy-4-fluorobenzene

  • MDL number MFCD00012201
  • Cas Number : 398-62-9
  • Molecular Weight : 156.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Fluoroveratrole
Purity 98
Density 1.17
Flash Point 91
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 120-123/45mm
Product Link https://store.apolloscientific.co.uk/product/12-dimethoxy-4-fluorobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/12-dimethoxy-4-fluorobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart