APO455835471 Apollo Scientific Ltd

3,5-Dibromo-4-hydroxybenzotrifluoride

  • MDL number MFCD09261146
  • Cas Number : 35852-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51-53
Synonym 2,6-Dibromo-4-(trifluoromethyl)phenol, 2,6-Dibromo-alpha,alpha,alpha-trifluoro-p-cresol
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=51295

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=51295
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart