APO455835472 Apollo Scientific Ltd

3-Amino-4-hydroxybenzotrifluoride

  • MDL number MFCD01731750
  • Cas Number : 454-81-9
  • Molecular Weight : 177.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118-120
Synonym 2-Hydroxy-5-(trifluoromethyl)aniline, 2-Amino-4-(trifluoromethyl)phenol, 6-Hydroxy-alpha,alpha,alpha-trifluoro-m-toluidine
Purity 95
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-amino-4-hydroxybenzotrifluoride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-4-hydroxybenzotrifluoride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart