APO455835474 Apollo Scientific Ltd

2-Methyl-6-(trifluoromethyl)-2H-1,4-benzoxazin-3(4H)-one

  • MDL number MFCD19376607
  • Cas Number : 189940-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3,4-Dihydro-2-methyl-3-oxo-6-(trifluoromethyl)-2H-1,4-benzoxazine, 2-Methyl-6-(trifluoromethyl)-4H-benzo[b][1,4]oxazin-3-one
Purity 95
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=51298

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=51298
Availability details for
Quantity
  • Availability
  • Add to Cart