APO455835478 Apollo Scientific Ltd

Methyl 2,4,5-trifluorobenzoate

  • MDL number MFCD06203702
  • Cas Number : 20372-66-1
  • Molecular Weight : 190.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Physical Form Solid-Fused
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 130-135/100mm
Product Link https://store.apolloscientific.co.uk/product/methyl-245-trifluorobenzoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-245-trifluorobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart