APO455835479 Apollo Scientific Ltd

2-Fluoro-4-(trifluoromethyl)benzeneboronic acid

  • MDL number MFCD03094995
  • Cas Number : 503309-11-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113-115
Synonym 4-Borono-3-fluorobenzotrifluoride
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=10380

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=10380
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart