APO455835480 Apollo Scientific Ltd

5-[1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]thiophene-2-sulphonyl chloride

  • MDL number MFCD00052283
  • Cas Number : 166964-31-4
  • Molecular Weight : 330.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77-80
Synonym 2-(Chlorosulphonyl)-5-[1-methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]thiophene, 3-[5-(Chlorosulphonyl)thien-2-yl]-1-methyl-5-(trifluoromethyl)-1H-pyrazole
Purity 97
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-ylthiophene-2-sulphonyl-chloride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-ylthiophene-2-sulphonyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart