APO455835483 Apollo Scientific Ltd

4-(Dimethylamino)-3-nitrobenzotrifluoride

  • MDL number MFCD00236670
  • Cas Number : 40700-38-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N,N-Dimethyl-2-nitro-4-(trifluoromethyl)aniline, 2-(Dimethylamino)-5-(trifluoromethyl)nitrobenzene
Purity 97
Flash Point >110
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=1516

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=1516
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart