APO455835490 Apollo Scientific Ltd

Dimethyl perfluorosebacate

  • MDL number MFCD00236604
  • Cas Number : 4590-24-3
  • Molecular Weight : 518.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Dimethyl hexadecafluorodecanedioate
Purity 96
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/dimethyl-perfluorosebacate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/dimethyl-perfluorosebacate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart