APO455835496 Apollo Scientific Ltd

3,5-Dinitrobenzotrifluoride

  • MDL number MFCD00007233
  • Cas Number : 401-99-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 47-52
Synonym 1,3-Dinitro-5-(trifluoromethyl)benzene, 3,5-Dinitro-alpha,alpha,alpha-trifluorotoluene
Purity 99
Flash Point >110
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 127-129/9mm
Product Link https://store.apolloscientific.co.uk/product/35-dinitrobenzotrifluoride-99

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/35-dinitrobenzotrifluoride-99
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart