APO455835502 Apollo Scientific Ltd

2,4-Dinitro-5-fluorotoluene

  • MDL number MFCD00205111
  • Cas Number : 349-01-9
  • Molecular Weight : 200.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 84-86
Synonym 2,4-Dinitro-1-fluoro-5-methylbenzene
Purity 99
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/24-dinitro-5-fluorotoluene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/24-dinitro-5-fluorotoluene
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart