APO455835509 Apollo Scientific Ltd

2,5-Bis(2,2,2-trifluoroethoxy)benzoic acid

  • MDL number MFCD00221424
  • Cas Number : 35480-52-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/25-bis222-trifluoroethoxybenzoic-acid-97

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/25-bis222-trifluoroethoxybenzoic-acid-97
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart