APO455835516 Apollo Scientific Ltd

Perfluoro-4-aminobiphenyl

  • MDL number MFCD00671483
  • Cas Number : 969-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145-146
Synonym 2,2',3,3',4',5,5',6,6'-Nonafluorobiphenyl-4-amine, 4-Amino-2,2',3,3',4',5,5',6,6'-nonafluorobiphenyl
Purity 96
Flash Point >110
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/perfluoro-4-aminobiphenyl-96

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/perfluoro-4-aminobiphenyl-96
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart