APO455835524 Apollo Scientific Ltd

2-Bromo-6-fluoropyridine

  • MDL number MFCD03095258
  • Cas Number : 144100-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 30-34
Purity 97
Density 1.71
Flash Point 75
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 162-164
Product Link https://store.apolloscientific.co.uk/product/2-bromo-6-fluoropyridine-97

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-6-fluoropyridine-97
pictograms GHS06
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart