APO455835525 Apollo Scientific Ltd

2-Chloro-6-fluoropyridine

  • MDL number MFCD00077483
  • Cas Number : 20885-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 34-36
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-chloro-6-fluoropyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-6-fluoropyridine
pictograms GHS07
hstatements H302+H312+H332;H319
Availability details for
Quantity
  • Availability
  • Add to Cart