APO455835529 Apollo Scientific Ltd

3,3'-Bis(trifluoromethyl)-[1,1'-biphenyl]-4,4'-diamine

  • MDL number MFCD00236672
  • Cas Number : 346-88-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116-118
Synonym 3,3'-Bis(trifluoromethyl)-4,4'-diaminobiphenyl, 3,3'-Bis(trifluoromethyl)benzidine
Purity 97
Flash Point 165
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=317

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=317
pictograms GHS06;GHS08;GHS09
hstatements H301+H311;H350;H330;H410
Availability details for
Quantity
  • Availability
  • Add to Cart