APO455835533 Apollo Scientific Ltd

2,4-Bis(trifluoromethyl)benzonitrile

  • MDL number MFCD00061225
  • Cas Number : 177952-38-4
  • Molecular Weight : 239.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/24-bistrifluoromethylbenzonitrile

Safety Information

signalword Danger
pcodes P301+P310
MSDS https://store.apolloscientific.co.uk/product/24-bistrifluoromethylbenzonitrile
pictograms GHS06
hstatements H301;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart