APO455835534 Apollo Scientific Ltd

2,6-Bis(trifluoromethyl)benzonitrile

  • MDL number MFCD00061226
  • Cas Number : 25753-25-7
  • Molecular Weight : 239.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,6-bis(trifluoromethyl)benzonitrile
Inchi Code 1S/C9H3F6N/c10-8(11,12)6-2-1-3-7(5(6)4-16)9(13,14)15/h1-3H
Inchi Key ANWBHDYEEOSTSE-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/26-bistrifluoromethylbenzonitrile

Safety Information

signalword Danger
pcodes P280
MSDS https://store.apolloscientific.co.uk/product/26-bistrifluoromethylbenzonitrile
pictograms GHS06
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart