APO455835536 Apollo Scientific Ltd

3,5-Bis(trifluoromethyl)benzonitrile

  • MDL number MFCD00000379
  • Cas Number : 27126-93-8
  • Molecular Weight : 239.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-21
Purity 97
Density 1.421
Flash Point 72
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 154-156
Product Link https://store.apolloscientific.co.uk/product/35-bistrifluoromethylbenzonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/35-bistrifluoromethylbenzonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart