APO455835538 Apollo Scientific Ltd

3,4'-Bis(trifluoromethyl)benzophenone

  • MDL number MFCD00013561
  • Cas Number : 21084-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93-97
Synonym [3-(Trifluoromethyl)phenyl][4-(trifluoromethyl)phenyl]methanone
Purity 98
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=327

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=327
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart