APO455835539 Apollo Scientific Ltd

3,5-Bis(trifluoromethyl)benzophenone

  • MDL number MFCD00015510
  • Cas Number : 21221-93-2
  • Molecular Weight : 318.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym [3,5-Bis(trifluoromethyl)phenyl](phenyl)methanone
Purity 97
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 111-112/0.2mm
Product Link https://store.apolloscientific.co.uk/product/35-bistrifluoromethylbenzophenone

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/35-bistrifluoromethylbenzophenone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart