APO455835540 Apollo Scientific Ltd

2,4-Bis(trifluoromethyl)benzoyl chloride

  • MDL number MFCD00061227
  • Cas Number : 53130-43-1
  • Molecular Weight : 276.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 179
Product Link https://store.apolloscientific.co.uk/product/24-bistrifluoromethylbenzoyl-chloride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/24-bistrifluoromethylbenzoyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart