APO455835543 Apollo Scientific Ltd

2,4-Bis(trifluoromethyl)benzyl alcohol

  • MDL number MFCD00042491
  • Cas Number : 143158-15-0
  • Molecular Weight : 244.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/24-bistrifluoromethylbenzyl-alcohol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/24-bistrifluoromethylbenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart