APO455835551 Apollo Scientific Ltd

3,5-Bis(trifluoromethyl)nitrobenzene

  • MDL number MFCD00000384
  • Cas Number : 328-75-6
  • Molecular Weight : 259.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1,3-Bis(trifluoromethyl)-5-nitrobenzene
Purity 98
Density 1.53
Flash Point 77
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 72/760mm
Product Link https://store.apolloscientific.co.uk/product/35-bistrifluoromethylnitrobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/35-bistrifluoromethylnitrobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart