APO455835552 Apollo Scientific Ltd

2-(1,3-Benzoxazol-2-yl)-1-[3-(trifluoromethyl)phenyl]ethan-1-ol

  • MDL number MFCD03424485
  • Cas Number : 502625-49-2
  • Molecular Weight : 307.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132-135
Synonym 2-(Benzo[d]oxazol-2-yl)-1-[3-(trifluoromethyl)phenyl]ethan-1-ol, 3-[2-(1,3-Benzoxazol-2-yl)-1-hydroxyethyl]benzotrifluoride
Purity 95
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-13-benzoxazol-2-yl-1-3-trifluoromethylphenylethan-1-ol

Safety Information

signalword Danger
pcodes P260;P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/2-13-benzoxazol-2-yl-1-3-trifluoromethylphenylethan-1-ol
pictograms GHS07;GHS08
hstatements H302+H312+H332;H315;H319;H334;H335
Availability details for
Quantity
  • Availability
  • Add to Cart