APO455835559 Apollo Scientific Ltd

3,5-Bis(trifluoromethyl)-DL-phenylalanine

  • MDL number MFCD00061309
  • Cas Number : 237076-69-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Amino-3-[3,5-bis(trifluoromethyl)phenyl]propanoic acid, 2-Amino-3-[3,5-bis(trifluoromethyl)phenyl]propionic acid
Purity 98
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=381

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=381
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart