APO455835564 Apollo Scientific Ltd

5,7-Bis(trifluoromethyl)-4-hydroxyquinoline

  • MDL number MFCD00153141
  • Cas Number : 237076-72-1
  • Molecular Weight : 281.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5,7-Bis(trifluoromethyl)quinolin-4-ol, 5,7-Bis(trifluoromethyl)-4-hydroxy-1-azanaphthalene
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/57-bistrifluoromethyl-4-hydroxyquinoline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/57-bistrifluoromethyl-4-hydroxyquinoline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart