APO455835574 Apollo Scientific Ltd

1H,1H,8H,8H-Dodecafluorooctane-1,8-diol

  • MDL number MFCD00153145
  • Cas Number : 90177-96-1
  • Molecular Weight : 362.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80-83
Synonym 2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluorooctane-1,8-diol, 1,8-Dihydroxy-2,2,3,3,4,4,5,5,6,6,7,7-dodecafluorooctane
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point >250
Product Link https://store.apolloscientific.co.uk/product/1h1h8h8h-dodecafluorooctane-18-diol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1h1h8h8h-dodecafluorooctane-18-diol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart