APO455835579 Apollo Scientific Ltd

1-(4-Fluorophenyl)-5-methyl-1H-pyrazole-4-carbonyl chloride

  • MDL number MFCD03659694
  • Cas Number : 423768-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90-92
Purity 97
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=6775

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=6775
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart