APO455835584 Apollo Scientific Ltd

(1S)-1-(3-Fluorophenyl)ethylamine

  • MDL number MFCD06761860
  • Cas Number : 444643-09-8
  • Molecular Weight : 139.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (S)-3-Fluoro-alpha-methylbenzylamine
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 63-67/4.2mm
Product Link https://store.apolloscientific.co.uk/product/1s-1-3-fluorophenylethylamine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1s-1-3-fluorophenylethylamine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart