APO455835587 Apollo Scientific Ltd

2-(Pyrazin-2-yl)-1-[4-(trifluoromethyl)phenyl]ethan-1-ol

  • MDL number MFCD03424491
  • Cas Number : 502625-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100-102
Synonym 2-(1,4-Diazin-2-yl)-1-[4-(trifluoromethyl)phenyl]ethan-1-ol, 4-[1-Hydroxy-2-(pyrazin-2-yl)ethyl]benzotrifluoride
Purity 95
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=6179

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=6179
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart