APO455835588 Apollo Scientific Ltd

1H,1H,11H-Icosafluoroundecan-1-ol

  • MDL number MFCD00039628
  • Cas Number : 307-70-0
  • Molecular Weight : 532.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95-97
Synonym 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-Icosafluoroundecan-1-ol, 1H,1H,11H-Perfluoroundecan-1-ol
Purity 85
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 180-181/200mm
Product Link https://store.apolloscientific.co.uk/product/1h1h11h-icosafluoroundecan-1-ol

Safety Information

signalword Warning
pcodes P280;P271;P261
MSDS https://store.apolloscientific.co.uk/product/1h1h11h-icosafluoroundecan-1-ol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart