APO455835594 Apollo Scientific Ltd

Ethyl 2-amino-4-(trifluoromethyl)-1,3-thiazole-5-carboxylate

  • MDL number MFCD00153147
  • Cas Number : 344-72-9
  • Molecular Weight : 240.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-172
Purity 97
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-2-amino-4-trifluoromethyl-13-thiazole-5-carboxylate

Safety Information

signalword Warning
pcodes P280;P301+P312;P302+P352
MSDS https://store.apolloscientific.co.uk/product/ethyl-2-amino-4-trifluoromethyl-13-thiazole-5-carboxylate
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart