APO455835597 Apollo Scientific Ltd

Ethyl 1,2-dihydro-2-oxo-6-(trifluoromethyl)pyrimidine-5-carboxylate

  • MDL number MFCD12547939
  • Cas Number : 154934-97-1
  • Molecular Weight : 236.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176-178
Synonym Ethyl 2-hydroxy-4-(trifluoromethyl)pyrimidine-5-carboxylate, Ethyl 1,2-dihydro-2-oxo-4-(trifluoromethyl)pyrimidine-5-carboxylate
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-12-dihydro-2-oxo-6-trifluoromethylpyrimidine-5-carboxylate

Safety Information

signalword Warning
pcodes P260;P280;P301+P312;P302+P352;P304+P340;P361+P364
MSDS https://store.apolloscientific.co.uk/product/ethyl-12-dihydro-2-oxo-6-trifluoromethylpyrimidine-5-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart