APO455835598 Apollo Scientific Ltd

3-Bromo-2-fluorobenzyl alcohol

  • MDL number MFCD09842441
  • Cas Number : 261723-32-4
  • Molecular Weight : 205.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (3-Bromo-2-fluorophenyl)methanol
Purity 98
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 104-108/4mm
Product Link https://store.apolloscientific.co.uk/product/3-bromo-2-fluorobenzyl-alcohol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-2-fluorobenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart