APO455835599 Apollo Scientific Ltd

3-Bromo-2-fluorobenzyl bromide

  • MDL number MFCD09842442
  • Cas Number : 149947-16-0
  • Molecular Weight : 267.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 60/0.1mm
Product Link https://store.apolloscientific.co.uk/product/3-bromo-2-fluorobenzyl-bromide

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-2-fluorobenzyl-bromide
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart