APO455835600 Apollo Scientific Ltd

Ethyl 6-chloro-5-cyano-2-(trifluoromethyl)nicotinate

  • MDL number MFCD00178758
  • Cas Number : 175277-73-3
  • Molecular Weight : 278.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78-81
Synonym 2-Chloro-5-(ethoxycarbonyl)-6-(trifluoromethyl)nicotinonitrile, Ethyl 6-chloro-5-cyano-2-(trifluoromethyl)pyridine-3-carboxylate
Purity 99
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-6-chloro-5-cyano-2-trifluoromethylnicotinate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-6-chloro-5-cyano-2-trifluoromethylnicotinate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart