APO455835603 Apollo Scientific Ltd

3-Chloro-2,4-difluoronitrobenzene

  • MDL number MFCD00042201
  • Cas Number : 3847-58-3
  • Molecular Weight : 193.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 45-49
Synonym 2-Chloro-1,3-difluoro-4-nitrobenzene
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloro-24-difluoronitrobenzene

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-24-difluoronitrobenzene
pictograms GHS05;GHS07;GHS09
hstatements H302;H314;H410
Availability details for
Quantity
  • Availability
  • Add to Cart