APO455835609 Apollo Scientific Ltd

Ethyl 1-(4-chlorophenyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD00052074
  • Cas Number : 112055-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 52-54
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 126-128/0.02mm
Product Link https://store.apolloscientific.co.uk/product/ethyl-1-4-chlorophenyl-5-trifluoromethyl-1h-pyrazole-4-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-1-4-chlorophenyl-5-trifluoromethyl-1h-pyrazole-4-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart