APO455835610 Apollo Scientific Ltd

2-Fluoro-6-[4-(methylthio)phenoxy]benzonitrile

  • MDL number MFCD00068069
  • Cas Number : 148901-52-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68-71
Synonym 2-Fluoro-6-[4-(methylsulphanyl)phenoxy]benzonitrile
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=7192

Safety Information

signalword Danger
pcodes P280;P301+P310;P304+P340
MSDS http://www.apolloscientific.co.uk/display_item.php?id=7192
pictograms GHS06
hstatements H301;H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart