APO455835612 Apollo Scientific Ltd

Methyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate

  • MDL number MFCD01570523
  • Cas Number : 655235-65-7
  • Molecular Weight : 239.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Methyl 3-chloro-5-(trifluoromethyl)picolinate, 3-Chloro-2-(methoxycarbonyl)-5-(trifluoromethyl)pyridine
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-3-chloro-5-trifluoromethylpyridine-2-carboxylate

Safety Information

signalword Warning
pcodes P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-3-chloro-5-trifluoromethylpyridine-2-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart