APO455835617 Apollo Scientific Ltd

4-Chloro-3-(trifluoromethyl)benzenesulphonyl chloride

  • MDL number MFCD00024882
  • Cas Number : 32333-53-2
  • Molecular Weight : 279.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-chloro-3-trifluoromethylbenzenesulphonyl-chloride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-3-trifluoromethylbenzenesulphonyl-chloride
pictograms GHS05
hstatements H290;H314
Availability details for
Quantity
  • Availability
  • Add to Cart