APO455835619 Apollo Scientific Ltd

Methyl 4-chloro-3-fluorobenzoate

  • MDL number MFCD00209299
  • Cas Number : 206362-87-0
  • Molecular Weight : 188.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-4-chloro-3-fluorobenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-4-chloro-3-fluorobenzoate
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart