APO455835627 Apollo Scientific Ltd

8-Fluoroquinoline

  • MDL number MFCD01685514
  • Cas Number : 394-68-3
  • Molecular Weight : 147.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.21
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 148-150/30mm
Product Link https://store.apolloscientific.co.uk/product/8-fluoroquinoline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/8-fluoroquinoline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart