APO455835628 Apollo Scientific Ltd

6-(2,2,2-Trifluoroethoxy)nicotinic acid

  • MDL number MFCD00110708
  • Cas Number : 175204-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174
Synonym 6-(2,2,2-Trifluoroethoxy)pyridine-3-carboxylic acid, 5-Carboxy-2-(2,2,2-trifluoroethoxy)pyridine
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/6-222-trifluoroethoxynicotinic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-222-trifluoroethoxynicotinic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart