APO455835634 Apollo Scientific Ltd

Ethyl 1-(2,4-difluorophenyl)-5-methylpyrazole-4-carboxylate

  • MDL number MFCD00067830
  • Cas Number : 175135-71-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 116-118/0.5mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=1590

Safety Information

signalword Warning
pcodes P280;P261
MSDS http://www.apolloscientific.co.uk/display_item.php?id=1590
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart