APO455835639 Apollo Scientific Ltd

3-Bromo-4-(trifluoromethoxy)aniline

  • MDL number MFCD00190127
  • Cas Number : 191602-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Flash Point >110
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-bromo-4-trifluoromethoxyaniline-98

Safety Information

signalword Warning
pcodes P280;P271;P260
MSDS https://store.apolloscientific.co.uk/product/3-bromo-4-trifluoromethoxyaniline-98
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart