APO455835647 Apollo Scientific Ltd

5-Oxo-1-[3-(trifluoromethyl)phenyl]pyrrolidine-3-carboxylic acid

  • MDL number MFCD00665765
  • Cas Number : 2357-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Carboxy-2-oxo-1-[3-(trifluoromethyl)phenyl]pyrrolidine, 3-(4-Carboxy-2-oxopyrrolidin-1-yl)benzotrifluoride
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4245

Safety Information

signalword Warning
pcodes P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4245
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart