APO455835649 Apollo Scientific Ltd

(2E)-1-(Fur-2-yl)-3-[4-(trifluoromethyl)phenyl]prop-2-en-1-one

  • MDL number MFCD00176655
  • Cas Number : 680215-11-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-[(1E)-3-(Fur-2-yl)-3-oxoprop-1-en-1-yl]benzotrifluoride, 2-{(2E)-3-[4-(Trifluoromethyl)phenyl]prop-2-enoyl}furan
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=7613

Safety Information

signalword Warning
pcodes P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=7613
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart